ENA185118574 Enamine

2-(4-fluorophenyl)-hexahydro-1H-pyrrolo[1,2-a]imidazolidine-1,3-dione

  • MDL number MFCD11857830
  • Cas Number : 60725-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41590

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41590
Availability details for
Quantity
  • Availability
  • Add to Cart