ENA185118579 Enamine

N-[4-(aminomethyl)phenyl]-4-phenoxybutanamide

  • MDL number MFCD09727952
  • Cas Number : 953898-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41596
Availability details for
Quantity
  • Availability
  • Add to Cart