ENA185118581 Enamine

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]pyrrolidine-2-carboxylic acid

  • MDL number MFCD07657733
  • Cas Number : 1188435-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41598

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41598
Availability details for
Quantity
  • Availability
  • Add to Cart