ENA185118587 Enamine

2-(bromomethyl)-4-fluoro-1-nitrobenzene

  • MDL number MFCD09736331
  • Cas Number : 82420-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41615
Availability details for
Quantity
  • Availability
  • Add to Cart