ENA185118593 Enamine

2-(2-methyl-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD11811769
  • Cas Number : 1171030-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41625
Availability details for
Quantity
  • Availability
  • Add to Cart