ENA185118596 Enamine

2-amino-5-propylbenzoic acid

  • MDL number MFCD11193635
  • Cas Number : 637347-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41629
Availability details for
Quantity
  • Availability
  • Add to Cart