ENA185118606 Enamine

N-[3-(aminomethyl)phenyl]butanamide

  • MDL number MFCD09807168
  • Cas Number : 918810-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41669
Availability details for
Quantity
  • Availability
  • Add to Cart