ENA185118607 Enamine

2-bromo-N-hydroxybenzamide

  • MDL number MFCD09940455
  • Cas Number : 2593-27-3
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41670
Availability details for
Quantity
  • Availability
  • Add to Cart