ENA185118617 Enamine

1-tert-butyl-3,6-dimethyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD11839876
  • Cas Number : 1118787-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41692
Availability details for
Quantity
  • Availability
  • Add to Cart