ENA185118620 Enamine

2-[(2-aminoethyl)amino]-6-fluorobenzonitrile hydrochloride

  • MDL number MFCD11798832
  • Cas Number : 1170539-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-41696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41696
Availability details for
Quantity
  • Availability
  • Add to Cart