ENA185118621 Enamine

1-methyl-5-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11122554
  • Cas Number : 706819-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41697
Availability details for
Quantity
  • Availability
  • Add to Cart