ENA185118629 Enamine

2-[2-(2,5-dimethyl-1H-pyrrol-1-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD09740169
  • Cas Number : 954564-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41722
Availability details for
Quantity
  • Availability
  • Add to Cart