ENA185118632 Enamine

4-(phenylcarbamoyl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

  • MDL number MFCD09728515
  • Cas Number : 953895-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41726
Availability details for
Quantity
  • Availability
  • Add to Cart