ENA185118635 Enamine

1-[6-(4-ethylpiperazin-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD09735019
  • Cas Number : 889851-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41729
Availability details for
Quantity
  • Availability
  • Add to Cart