ENA185118639 Enamine

N-(piperidin-4-yl)butanamide hydrochloride

  • MDL number MFCD11811767
  • Cas Number : 71879-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40200
Availability details for
Quantity
  • Availability
  • Add to Cart