ENA185118655 Enamine

2-(4-chlorophenyl)-2,2-difluoroacetic acid

  • MDL number MFCD11007727
  • Cas Number : 475301-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41782
Availability details for
Quantity
  • Availability
  • Add to Cart