ENA185118658 Enamine

5-(chloromethyl)-N,N-dimethylfuran-2-sulfonamide

  • MDL number MFCD11840108
  • Cas Number : 56038-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41787
Availability details for
Quantity
  • Availability
  • Add to Cart