ENA185118662 Enamine

2-nitro-5-(propan-2-yloxy)benzoic acid

  • MDL number MFCD09814881
  • Cas Number : 78361-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41792
Availability details for
Quantity
  • Availability
  • Add to Cart