ENA185118664 Enamine

5-[(4-bromo-3-methylphenyl)methyl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD11795704
  • Cas Number : 1171901-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41794
Availability details for
Quantity
  • Availability
  • Add to Cart