ENA185118665 Enamine

4-(2,3-dihydro-1H-indol-1-yl)aniline

  • MDL number MFCD08699237
  • Cas Number : 224036-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41795
Availability details for
Quantity
  • Availability
  • Add to Cart