ENA185118667 Enamine

5-chloro-1,3-dimethyl-4-nitro-1H-pyrazole

  • MDL number MFCD00052534
  • Cas Number : 13551-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78...80
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41798

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-41798
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart