ENA185118668 Enamine

2-(4-aminopiperidin-1-yl)-N-(propan-2-yl)acetamide dihydrochloride

  • MDL number MFCD11802523
  • Cas Number : 1171796-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41799
Availability details for
Quantity
  • Availability
  • Add to Cart