ENA185118677 Enamine

2-methyl-N-(piperidin-4-yl)propanamide hydrochloride

  • MDL number MFCD11809618
  • Cas Number : 1170578-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40201
Availability details for
Quantity
  • Availability
  • Add to Cart