ENA185118686 Enamine

1-(4-aminopiperidin-1-yl)-3-phenylpropan-1-one hydrochloride

  • MDL number MFCD11802849
  • Cas Number : 1158369-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41835
Availability details for
Quantity
  • Availability
  • Add to Cart