ENA185118688 Enamine

3-amino-4-methyl-N-phenylbenzamide

  • MDL number MFCD09050705
  • Cas Number : 54884-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41837
Availability details for
Quantity
  • Availability
  • Add to Cart