ENA185118690 Enamine

4-fluoro-3-[(1H-imidazol-1-yl)methyl]benzonitrile

  • MDL number MFCD09936347
  • Cas Number : 1016874-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41839
Availability details for
Quantity
  • Availability
  • Add to Cart