ENA185118691 Enamine

5-[(3,4-dimethylphenyl)methyl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD11795703
  • Cas Number : 1171575-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41840
Availability details for
Quantity
  • Availability
  • Add to Cart