ENA185118692 Enamine

2-({[(4-bromophenyl)carbamoyl]methane}sulfonyl)acetic acid

  • MDL number MFCD02597734
  • Cas Number : 1016777-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41841
Availability details for
Quantity
  • Availability
  • Add to Cart