ENA185118693 Enamine

2-(2-amino-4-fluorophenyl)-octahydro-1H-isoindole-1,3-dione

  • MDL number MFCD09741752
  • Cas Number : 954582-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41842
Availability details for
Quantity
  • Availability
  • Add to Cart