ENA185118709 Enamine

5-(morpholine-4-sulfonyl)thiophene-3-carboxylic acid

  • MDL number MFCD11122624
  • Cas Number : 1041528-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41869
Availability details for
Quantity
  • Availability
  • Add to Cart