ENA185118716 Enamine

2-chloro-N-hexylacetamide

  • MDL number MFCD00085727
  • Cas Number : 5326-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29...31
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23385

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23385
Availability details for
Quantity
  • Availability
  • Add to Cart