ENA185118721 Enamine

1-[2-(3,4-dimethylphenoxy)pyridin-4-yl]methanamine hydrochloride

  • MDL number MFCD11802869
  • Cas Number : 1172735-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41890
Availability details for
Quantity
  • Availability
  • Add to Cart