ENA185118724 Enamine

6-ethoxy-2H-chromene-3-carboxylic acid

  • MDL number MFCD08444974
  • Cas Number : 923191-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41893
Availability details for
Quantity
  • Availability
  • Add to Cart