ENA185118729 Enamine

methyl 5-amino-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD11053056
  • Cas Number : 222970-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41900
Availability details for
Quantity
  • Availability
  • Add to Cart