ENA185118738 Enamine

6-(4-fluorophenyl)-4-methyl-1H,6H-[1,2]diazolo[3,4-c]pyrazol-3-amine

  • MDL number MFCD09042032
  • Cas Number : 926270-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41938
Availability details for
Quantity
  • Availability
  • Add to Cart