ENA185118747 Enamine

3-(piperazin-1-yl)-6-(1H-pyrazol-1-yl)pyridazine

  • MDL number MFCD11814078
  • Cas Number : 1172883-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41955
Availability details for
Quantity
  • Availability
  • Add to Cart