ENA185118748 Enamine

4-(2,4-dimethyl-1,3-thiazol-5-yl)-1,3-thiazol-2-amine

  • MDL number MFCD00775052
  • Cas Number : 7520-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41956

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41956
Availability details for
Quantity
  • Availability
  • Add to Cart