ENA185118752 Enamine

(4-amino-3-nitrophenyl)(phenyl)methanol

  • MDL number MFCD08442155
  • Cas Number : 115577-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41960
Availability details for
Quantity
  • Availability
  • Add to Cart