ENA185118755 Enamine

methyl 2-{[4-(propan-2-yl)phenyl]sulfanyl}acetate

  • MDL number MFCD11184358
  • Cas Number : 875256-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41965
Availability details for
Quantity
  • Availability
  • Add to Cart