ENA185118759 Enamine

6-[(2-aminoethyl)amino]pyridine-3-sulfonamide

  • MDL number MFCD09041001
  • Cas Number : 202460-51-3
  • Molecular Weight : 216.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41213
Availability details for
Quantity
  • Availability
  • Add to Cart