ENA185118761 Enamine

N-(2-nitrophenyl)pyrrolidine-1-carboxamide

  • MDL number MFCD11728278
  • Cas Number : 1090012-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41982
Availability details for
Quantity
  • Availability
  • Add to Cart