ENA185118768 Enamine

1lambda6-benzothiophene-1,1-dione

  • MDL number MFCD00052178
  • Cas Number : 825-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-143
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42002

Safety Information

signalword Danger
pcodes P261,P264,P270,P272,P280,P301+P310,P302+P352,P330,P333+P313,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-42002
pictograms GHS06
hstatements H301,H317
Availability details for
Quantity
  • Availability
  • Add to Cart