ENA185118774 Enamine

3-(3-chloro-4-methylphenyl)-4-oxo-3,4-dihydrophthalazine-1-carboxylic acid

  • MDL number MFCD09048651
  • Cas Number : 926201-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42010
Availability details for
Quantity
  • Availability
  • Add to Cart