ENA185118775 Enamine

ethyl[(oxolan-3-yl)methyl]amine

  • MDL number MFCD09054805
  • Cas Number : 165253-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42012
Availability details for
Quantity
  • Availability
  • Add to Cart