ENA185118794 Enamine

4-tert-butyl 2-ethyl 5-(2-chloroacetamido)-3-methylthiophene-2,4-dicarboxylate

  • MDL number MFCD11839804
  • Cas Number : 1118787-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41300

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41300
Availability details for
Quantity
  • Availability
  • Add to Cart