ENA185118804 Enamine

2-ethyl-1-[4-(hydroxyimino)piperidin-1-yl]butan-1-one

  • MDL number MFCD09938072
  • Cas Number : 1016867-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41408
Availability details for
Quantity
  • Availability
  • Add to Cart