ENA185118807 Enamine

3-(3-ethoxyphenyl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD11506472
  • Cas Number : 1187631-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41463
Availability details for
Quantity
  • Availability
  • Add to Cart