ENA187622728 Enamine

4-(4,5,6,7-tetrahydro-1H-indazol-3-yl)aniline

  • MDL number MFCD11857818
  • Cas Number : 1118787-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41446
Availability details for
Quantity
  • Availability
  • Add to Cart