ENA187622729 Enamine

(4,5-dimethyl-4H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD11857828
  • Cas Number : 881845-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41584

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41584
Availability details for
Quantity
  • Availability
  • Add to Cart