ENA187622737 Enamine

1,4-dichloro-2-(2-chloroethanesulfinyl)benzene

  • MDL number MFCD11857870
  • Cas Number : 1172485-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-42138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42138
Availability details for
Quantity
  • Availability
  • Add to Cart